Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HM74A/PUMA-G/GPR109A/NIACR1 Antibody (MM0329-6S24), PE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212358PE
Description
HM74A/PUMA-G/GPR109A/NIACR1 Monoclonal specifically detects HM74A/PUMA-G/GPR109A/NIACR1 in Human samples. It is validated for Flow Cytometry.Specifications
HM74A/PUMA-G/GPR109A/NIACR1 | |
Monoclonal | |
PE | |
Q8TDS4.1 | |
HCAR2 | |
Human recombinant HM74A/PUMA-G/GPR109A/NIACR1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
MM0329-6S24 | |
Flow Cytometry | |
GPR109A, HCA2, hydroxycarboxylic acid receptor 2, NIACR1 | |
Rat | |
Protein G purified | |
RUO | |
338442 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction