Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMBOX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565983
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565983 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565983 Supplier Invitrogen™ Supplier No. PA565983
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IKRRANIEAAI LESHGIDVQS PGGHSNSDDV DGNDYSEQDD STSHSDHQDP ISLAVEMAAV NHTILALARQ GANEIKTEAL DD Highest antigen sequence identity to the following orthologs - mouse 100%, rat 47%.

HMBOX1 is located at the boundary of 8p12.3 and 8p21.1. HMBOX1 proteins are highly conserved in human, mouse, rat, chicken and Xenopus laevis. Functional HMBOX1::EGFP (enhanced green fluorescent protein) fusion protein revealed that HMBOX1 accumulated more in cytoplasm than in nucleus. HMBOX1 is a transcription repressor. Human HMBOX1 is widely expressed in 18 tissues, and it is highly expressed in pancreas. Hmbox1 is widely expressed in mouse pancreas and the expression of this gene can also be detected in pallium, hippocampus and hypothalamus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMBOX1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HMBOX1
Gene Accession No. Q6NT76
Gene Alias AI451877; AI604847; F830020C16Rik; HMBOX1; HNF1LA; homeobox containing 1; homeobox telomere-binding protein 1; Homeobox-containing protein 1; homeobox-containing protein PBHNF; HOT1; PBHNF; RGD1306787; TAH1; Telomere-associated homeobox-containing protein 1
Gene Symbols HMBOX1
Host Species Rabbit
Immunogen Recombinant Human HMBOX1. Recombinant protein control fragment (Product #RP-106670).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79618
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.