Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMBS Antibody (OTI3F4), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245976
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
HMBS Monoclonal specifically detects HMBS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
HMBS | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
hydroxymethylbilane synthasePre-uroporphyrinogen synthase, PBGDPBG-D, PORC, porphobilinogen deaminase, porphyria, acute; Chester type, UPSEC 2.5.1.61, uroporphyrinogen I synthase, uroporphyrinogen I synthetase | |
Mouse | |
39.1 kDa | |
0.1 mL | |
Diabetes Research | |
3145 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI3F4 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
HMBS | |
Full length human recombinant protein of human HMBS (NP_000181) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction