Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMBS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595924 Supplier Invitrogen™ Supplier No. PA595924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, Raji, Mouse spleen; Cellular Location: Cytoplasm.

PBGD (porphobilinogen deaminase), also designated hydroxymethylbilane synthase, is a cytoplasmic enzyme found in the heme synthesis pathway. PBGD belongs to the HMBS (hydroxymethylbilane synthase) family. Deficiency of PBGD causes errors in pyrrole metabolism, which in turn leads to an inherited autosomal disorder called acute intermittent porphyria (AIP). AIP is characterized by acute attacks of neurological dysfunctions with hypertension, tachycardia, peripheral neurologic disturbances, abdominal pain and excessive amounts of aminolevulinic acid and porphobilinogen in the urine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMBS
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 6.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Hmbs
Gene Accession No. P08397, P22907
Gene Alias alternative name: porphobilinogen deaminase; HEM3; hemC; HMBS; Hydroxymethylbilane synthase; PBG D; PBGD; PBG-D; PORC; Porphobilinogen deaminase; porphyria, acute; Chester type; pre-uroporphyrinogen synthase; similar to Porphobilinogen deaminase (Hydroxymethylbilane synthase) (HMBS) (Pre-uroporphyrinogen synthase) (PBG-D); T25658; Ups; uroporphyrinogen I synthase; uroporphyrinogen I synthetase; URO-S; Uros1
Gene Symbols Hmbs
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15288, 3145
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.