Learn More
HMBS Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304234AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
PBGD (porphobilinogen deaminase), also designated hydroxymethylbilane synthase, is a cytoplasmic enzyme found in the heme synthesis pathway. PBGD belongs to the HMBS (hydroxymethylbilane synthase) family. Deficiency of PBGD causes errors in pyrrole metabolism, which in turn leads to an inherited autosomal disorder called acute intermittent porphyria (AIP). AIP is characterized by acute attacks of neurological dysfunctions with hypertension, tachycardia, peripheral neurologic disturbances, abdominal pain and excessive amounts of aminolevulinic acid and porphobilinogen in the urine.Specifications
HMBS | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P08397 | |
HMBS | |
Between 311 and 361 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
HMBS | |
HEM3, HMBS, Hydroxymethylbilane synthase, PBG D, PBG-D, PBGD, Porphobilinogen deaminase, Pre uroporphyrinogen synthase, Pre-uroporphyrinogen synthase, UPS, Uroporphyrinogen I synthase, Uroporphyrinogen I synthetase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3145 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.