Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMCES Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318456
This item is not returnable.
View return policy
Description
HMCES Polyclonal antibody specifically detects HMCES in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
HMCES | |
Polyclonal | |
Unconjugated | |
Tris-citrate/phosphate pH 7 -8. | |
Rabbit | |
Affinity purified | |
RUO | |
56941 | |
Store at 4°C. Do not freeze. | |
IgG |
Western Blot, Immunoprecipitation | |
1 mg/ml | |
Western Blot 1:2000 - 1:10000, Immunoprecipitation 40 ug/mg lysate | |
chromosome 3 open reading frame 37, DC12, MGC111075 | |
Recognizes region between residues Between Between 304 and 354.. | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction