Learn More
Invitrogen™ HMCES Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 79%; Rabbit: 100%; Rat: 93%.
Specifications
Specifications
| Antigen | HMCES |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | HMCES |
| Gene Accession No. | Q8R1M0 |
| Gene Alias | 5-hydroxymethylcytosine (hmC) binding, ES cell specific; 5-hydroxymethylcytosine (hmC) binding, ES cell-specific; 5-hydroxymethylcytosine binding, ES cell specific; 8430410A17Rik; Abasic site processing protein HMCES; abasic site processing protein HMCES; embryonic stem cell-specific 5-hydroxymethylcytosine-binding protein; C20H3orf37; C22H3orf37; C3orf37; C85376; DC12; Embryonic stem cell-specific 5-hydroxymethylcytosine-binding protein; ES cell-specific 5hmC-binding protein; HMCES; Peptidase HMCES; Putative endonuclease HMCES; putative peptidase SRAPD1; SRAP domain-containing protein 1; Srap1; Srapd1; UPF0361 protein C3orf37; UPF0361 protein C3orf37 homolog |
| Gene Symbols | HMCES |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.