Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMG-CoA Reductase/HMGCR Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30377820UL
Description
HMG-CoA Reductase/HMGCR Polyclonal specifically detects HMG-CoA Reductase/HMGCR in Human, Mouse samples. It is validated for Western Blot, Knockout Validated.Specifications
HMG-CoA Reductase/HMGCR | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
3156 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, KnockDown | |
Unconjugated | |
3-hydroxy-3-methylglutaryl CoA reductase (NADPH), 3-hydroxy-3-methylglutaryl-CoA reductase, 3-hydroxy-3-methylglutaryl-Coenzyme A reductase, EC 1.1.1, EC 1.1.1.34, HMG-CoA reductase, hydroxymethylglutaryl-CoA reductase, LDLCQ3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 629-888 of human HMG-CoA Reductase/HMGCR (NP_000850.1). | |
20 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction