Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMG-CoA Reductase/HMGCR Rabbit anti-Human, Mouse, Rat, Clone: 6Y9L8, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315641100UL
Description
HMG-CoA Reductase/HMGCR Monoclonal antibody specifically detects HMG-CoA Reductase/HMGCR in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
HMG-CoA Reductase/HMGCR | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
6Y9L8 | |
Western Blot 1:500 - 1:2000 | |
3-hydroxy-3-methylglutaryl CoA reductase (NADPH), 3-hydroxy-3-methylglutaryl-CoA reductase, 3-hydroxy-3-methylglutaryl-Coenzyme A reductase, EC 1.1.1, EC 1.1.1.34, HMG-CoA reductase, hydroxymethylglutaryl-CoA reductase, LDLCQ3 | |
A synthetic peptide corresponding to a sequence within amino acids 400-500 of human HMG-CoA Reductase/HMGCR (P04035). IKPLVAETDTPNRATFVVGNSSLLDTSSVLVTQEPEIELPREPRPNEECLQILGNAEKGAKFLSDAEIIQLVNAKHIPAYKLETLMETHERGVSIRRQLLS | |
100 μg | |
Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
3156 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction