Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMG-Lysine Rabbit anti-All, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABS2108100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS2108-100 100 μL
ABS2108-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABS2108-100 Supplier MilliporeSigma Supplier No. ABS2108100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-HMG-Lysine, Cat. No. ABS2108, is a rabbit polyclonal antibody that detects hydroxymethylglutaryl-lysine conjugated proteins and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMG-Lysine
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in PBS with 0.1% sodium azide.
Gene Alias 3-hydroxy-3-methyl-glutary-Lysine
Host Species Rabbit
Immunogen 3-hydroxy-3-methyl-glutaryl (HMG)-Lysine conjugated to BSA.
Molecular Weight of Antigen ∽75 kDa observed (BSA conjugate). Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects BSA-conjugated 3-hydroxy-3-methyl-glutaryl (HMG)-Lysine. It recognizes BSA chemically modified to contain HMG-lysine modifications, but not unmodified BSA.
Target Species All species
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.