Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMG1L10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$464.00
Specifications
Antigen | HMG1L10 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
HMG1L10 Polyclonal specifically detects HMG1L10 in Human samples. It is validated for Western Blot.Specifications
HMG1L10 | |
Polyclonal | |
Purified | |
RUO | |
bK445C9.3, HMG1L10, HMGB1L10, HMGB1P10 high mobility group box 1 pseudogene 10 | |
HMGB1P10 | |
IgG | |
Protein A purified | |
23 kDa |
Western Blot | |
Unconjugated | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
100130561 | |
Synthetic peptide directed towards the C terminal of human HMG1L10. Peptide sequence YEKDIAAYRAKGKPDAAKKGVVKAEKSKKKKEEEEDEEDEEDEEEEDEED. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title