Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMG20A Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB300860
Description
HMG20A Polyclonal specifically detects HMG20A in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
HMG20A | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
FLJ10739, high-mobility group 20A, HMG box domain containing 1, HMG box-containing protein 20A, HMG domain-containing protein 1, HMGX1HMG domain-containing protein HMGX1, HMGXB1high mobility group protein 20A | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Cross reacts with Human and Rat. Expected from sequence similarity: Mouse, Dog, Pig and Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_060670. | |
HMG20A | |
Peptide with sequence C-ATVREVVNRLDR corresponding to C-Terminus according to NP_060670.1. | |
0.1 mg | |
Chromatin Research, Stem Cell Markers | |
10363 | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction