Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMG20A Antibody (OTI2H4), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271596AF405
Description
HMG20A Monoclonal specifically detects HMG20A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
HMG20A | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
HMG20A | |
Recombinant protein fragment corresponding to amino acids 1-281 of human HMG20A (NP_060670) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2H4 | |
Western Blot, Immunohistochemistry | |
FLJ10739, high-mobility group 20A, HMG box domain containing 1, HMG box-containing protein 20A, HMG domain-containing protein 1, HMGX1HMG domain-containing protein HMGX1, HMGXB1high mobility group protein 20A | |
Mouse | |
Affinity Purified | |
Chromatin Research, Stem Cell Markers | |
10363.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction