Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMG20A Monoclonal Antibody (OTI5E3)

HMG20A Monoclonal Antibody (OTI5E3)

Supplier:  Thermo Scientific MA526832

Catalog No. PIMA526832


Only null left
Add to Cart

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HMG20A
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NP66
HMG20A
Human recombinant protein fragment corresponding to amino acids 1-281 of HMG20A produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI5E3
Unconjugated
HMG20A
1200004E06Rik; 5730490E10Rik; FLJ10739; high mobility group 20A; high mobility group protein 20A; HMG box domain containing 1; HMG box-containing protein 20A; HMG domain-containing protein 1; HMG domain-containing protein HMGX1; HMG20A; HMGX1; HMGXB1; Ibraf; Inhibitor of BRAF35; RGD1564760
Mouse
Affinity Chromatography
RUO
10363
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.