Learn More
HMG2a Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | HMG2a |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1620
![]() |
Bethyl Laboratories, Inc
IHC00173T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1619
![]() |
Bethyl Laboratories, Inc
IHC00173 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan, Monkey, Gorilla, Chimpan
HMGB3 (HMG4) belongs to the high mobility group protein superfamily. Like HMG1 and HMG2 , HMGB3 contains DNA-binding HMG box domains and is classified into the HMG box subfamily. Members of the HMG box subfamily are thought to play a fundamental role in DNA replication, nucleosome assembly and transcription .Specifications
HMG2a | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
high mobility group protein 2a, High mobility group protein 4, high-mobility group (nonhistone chromosomal) protein 4, HMG2A, HMG-2a, HMG4, HMG-4, MGC90319, non-histone chromosomal protein | |
Between 125 and 175 | |
Primary | |
4° C | |
HMGB3 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O15347, O54879 | |
15354, 3149 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.