Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMG2L1 Antibody (OTI1H10), Biotin, Novus Biologicals™
SDP

Catalog No. NBP271538B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271538B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271538B Supplier Novus Biologicals Supplier No. NBP271538B
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HMG2L1 Monoclonal specifically detects HMG2L1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen HMG2L1
Applications Western Blot
Classification Monoclonal
Clone OTI1H10
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias High mobility group protein 2-like 1, high-mobility group protein 2-like 1, HMG box domain containing 4, HMG domain-containing protein 4, HMG2L1HMG box-containing protein 4, HMGBCG, Protein HMGBCG, THC211630
Gene Symbols HMGXB4
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-348 of human HMG2L1 (NP_005478) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 10042.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.