Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGA1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP32131825UL
Description
HMGA1 Polyclonal antibody specifically detects HMGA1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
HMGA1 | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
high mobility group AT-hook 1, High mobility group AT-hook protein 1, High mobility group protein A1, high mobility group protein HMG-I/HMG-Y, High mobility group protein R, high-mobility group (nonhistone chromosomal) protein isoforms I and Y, HMGA1A, HMG-I(Y), HMGIYMGC12816, HMG-R, MGC4242, MGC4854, nonhistone chromosomal high-mobility group protein HMG-I/HMG-Y | |
This antibody was developed against Recombinant Protein corresponding to amino acids: PKGSKNKGAAKTRKTTTTPGRKPRGRPKKLEKEE | |
25 μg | |
Cell Cycle and Replication | |
3159 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction