Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMGA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519184
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519184 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519184 Supplier Invitrogen™ Supplier No. PA519184
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.

HMGA1 encodes a non-histone protein involved in many cellular processes, including regulation of inducible gene transcription, integration of retroviruses into chromosomes, and the metastatic progression of cancer cells. The encoded protein preferentially binds to the minor groove of A+T-rich regions in double-stranded DNA. It has little secondary structure in solution but assumes distinct conformations when bound to substrates such as DNA or other proteins. The encoded protein is frequently acetylated and is found in the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene HMGA1
Gene Accession No. P17096
Gene Alias AL023995; HGNC:5010; high mobility group AT-hook 1; high mobility group AT-hook protein 1; high mobility group HMGA1A; high mobility group HMGA1B; high mobility group protein; high mobility group protein 1A; high mobility group protein A1; high mobility group protein A1A; high mobility group protein A1B; high mobility group protein HMG-I/HMG-Y; high mobility group protein I; high mobility group protein R; high mobiliy group protein A1B; high-mobility group (nonhistone chromosomal) protein isoforms I and Y; HMGA1; Hmga1a; Hmga1b; Hmgi; HMG-I(Y); HMGIY; HMG-R; Hmgy; MGC12816; MGC4242; MGC4854; nonhistone chromosomal high-mobility group protein HMG-I/HMG-Y; non-histone protein; non-histone protein; HMGA1A; non-histone protein; HMGA1B; OTTHUMP00000016222; OTTHUMP00000016223; OTTHUMP00000016224; OTTHUMP00000039618
Gene Symbols HMGA1
Host Species Goat
Immunogen Synthetic peptide sequence (SQPLASKQEKDGT) corresponding to the internal amino acids of HMGA1 (aa 9-22).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3159
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.