Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGA1 Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$156.65 - $620.58
Specifications
Antigen | HMGA1 |
---|---|
Concentration | Conc. Not Determined |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-4078
![]() ![]() |
Active Motif
39650 |
10 μL |
Each for $156.65
|
|
|||||
50-199-4077
![]() ![]() |
Active Motif
39615 |
100 μL |
Each for $620.58
|
|
|||||
Description
HMGA1 encodes a non-histone protein involved in many cellular processes, including regulation of inducible gene transcription, integration of retroviruses into chromosomes, and the metastatic progression of cancer cells. The encoded protein preferentially binds to the minor groove of A+T-rich regions in double-stranded DNA. It has little secondary structure in solution but assumes distinct conformations when bound to substrates such as DNA or other proteins. The encoded protein is frequently acetylated and is found in the nucleus.Specifications
HMGA1 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P17096 | |
3159 | |
This HMGA1 antibody was raised against a peptide corresponding to the N-terminus of human HMGA1. | |
Primary | |
Antibody |
Conc. Not Determined | |
Polyclonal | |
Liquid | |
RUO | |
whole serum with 30% glycerol and 0.035% sodium azide; pH | |
AL023995; HGNC:5010; high mobility group AT-hook 1; High mobility group AT-hook protein 1; high mobility group HMGA1A; high mobility group HMGA1B; high mobility group protein 1A; High mobility group protein A1; High mobility group protein HMG-I/HMG-Y; high mobility group protein I; High mobility group protein R; high-mobility group (nonhistone chromosomal) protein isoforms I and Y; Hmga1; Hmga1a; Hmga1b; Hmgi; HMG-I(Y); Hmgiy; HMG-R; Hmgy; MGC12816; MGC4242; MGC4854; nonhistone chromosomal high-mobility group protein HMG-I/HMG-Y; non-histone protein; OTTHUMP00000016222; OTTHUMP00000016223; OTTHUMP00000016224; OTTHUMP00000039618 | |
HMGA1 | |
IgG | |
-20°C, Avoid Freeze/Thaw Cycles | |
HMGA1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title