Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGA2 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298735H
Description
HMGA2 Polyclonal specifically detects HMGA2 in Human samples. It is validated for Western Blot.Specifications
HMGA2 | |
Polyclonal | |
Western Blot | |
BABLHMGI-C, high mobility group AT-hook 2, High mobility group AT-hook protein 2, high mobility group protein HMGI-C, high-mobility group (nonhistone chromosomal) protein isoform I-C, High-mobility group protein HMGI-C, HMGICSTQTL9, LIPO | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human HMGA2. | |
0.1 mL | |
Cancer | |
8091 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
HRP | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction