Learn More
HMGB1/HMG-1 Antibody (19N10B7), Alexa Fluor™ 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP227395AF350
Catalog No. NP22739A30
Only null left
Add to Cart
Description
Description
HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
HMGB1/HMG-1
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
HMGB1
Full-length recombinant human HMGB1 protein
Primary
This antibody recognizes both A box domain (amino acids 9-79) and full length HMGB1 protein.
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
19N10B7
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Mouse
0.1 mL
3146
Human, Mouse
IgG2b κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.