Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGB1/HMG-1 Antibody (19N10B7) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$192.94 - $519.67
Specifications
| Antigen | HMGB1/HMG-1 |
|---|---|
| Clone | 19N10B7 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-3 ug/ml. Use reported by customer review, Simple Western 1:50, Flow Cytometry 0.5ug/ 5x10^5cells, Immunohistochemistry 5 ug/ml, Immunoprecipitation reported in scientific literature (PMID 23696858), Immunohistochemistry-Paraffin 5 ug/ml, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227396SS
![]() |
Novus Biologicals
NBP227396SS |
0.025 mg |
Each for $192.94
|
|
|||||
NBP227396
![]() |
Novus Biologicals
NBP227396 |
0.1 mg |
Each for $519.67
|
|
|||||
Description
HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| HMGB1/HMG-1 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators | |
| P09429 | |
| 3146 | |
| Full-length recombinant human HMGB1 protein | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 19N10B7 | |
| Western Blot 1-3 ug/ml. Use reported by customer review, Simple Western 1:50, Flow Cytometry 0.5ug/ 5x10^5cells, Immunohistochemistry 5 ug/ml, Immunoprecipitation reported in scientific literature (PMID 23696858), Immunohistochemistry-Paraffin 5 ug/ml, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
| HMGB1 | |
| IgG2b κ | |
| Protein G purified | |
| This antibody recognizes both A box domain (amino acids 9-79) and full length HMGB1 protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title