Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGB1/HMG-1 Antibody (19N12A1), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227400AF488
Description
HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Western Blot.Specifications
HMGB1/HMG-1 | |
Monoclonal | |
Alexa Fluor 488 | |
P09429 | |
HMGB1 | |
Full-length recombinant human HMGB1 was used as the immunogen. | |
0.1 mL | |
Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators | |
3146 | |
Human, Mouse | |
Purified |
Western Blot | |
19N12A1 | |
Western Blot | |
Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This antibody recognizes both B box domain (amino acids 88-162) and full length HMGB1 protein. | |
Store at 4C in the dark. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction