Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGB1/HMG-1 Antibody (19N12A1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$206.50 - $501.50
Specifications
Antigen | HMGB1/HMG-1 |
---|---|
Clone | 19N12A1 |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1 - 3 ug/mL, Simple Western 1:100, Flow Cytometry reported in scientific literature (PMID 29601597) |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227400SS
![]() |
Novus Biologicals
NBP227400SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227400
![]() |
Novus Biologicals
NBP227400 |
0.1 mg |
Each for $501.50
|
|
|||||
Description
HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry.Specifications
HMGB1/HMG-1 | |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators | |
PBS with 0.05% Sodium Azide | |
Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
HMGB1 | |
IgG2b κ | |
Protein G purified | |
This antibody recognizes both B box domain (amino acids 88-162) and full length HMGB1 protein. |
19N12A1 | |
Western Blot 1 - 3 ug/mL, Simple Western 1:100, Flow Cytometry reported in scientific literature (PMID 29601597) | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
P09429 | |
3146 | |
Full-length recombinant human HMGB1 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title