Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGB1/HMG-1 Antibody (19N15F4) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280775
Description
HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.Specifications
HMGB1/HMG-1 | |
Monoclonal | |
0.5 mg/mL | |
Western Blot : 1-3 μg/mL, Flow Cytometry 5x10^5cells, Immunohistochemistry, Immunohistochemistry-Paraffin : 5 μg/mL, Immunocytochemistry | |
Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
Mouse | |
Protein G purified | |
RUO | |
3146 | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
19N15F4 | |
Unconjugated | |
PBS | |
HMGB1 | |
Full-length recombinant human HMGB1 was used as the immunogen for the antibody. | |
0.1 mg | |
Primary | |
Based upon immunogen sequence similarity, this antibody is predicted to react 100% with olive baboon, crab-eating macaque, and common marmoset. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction