Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGB1/HMG-1 Antibody (19N15F4), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227401FR
Description
HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HMGB1/HMG-1 | |
Monoclonal | |
DyLight 680 | |
P09429 | |
HMGB1 | |
Full-length recombinant human HMGB1 was used as the immunogen for the antibody. | |
0.1 mL | |
Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators | |
3146 | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
19N15F4 | |
Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin | |
Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This antibody recognizes both the box A domain (amino acids 9-79) and full length HMGB1 protein. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction