Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGB1/HMG-1 Antibody (19N15F4), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $426.00
Specifications
Antigen | HMGB1/HMG-1 |
---|---|
Clone | 19N15F4 |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1-3ug/ml∼, Flow Cytometry 0.5ug/5x10^5cells∼, Immunohistochemistry, Immunohistochemistry-Paraffin 5ug/ml |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227401SS
![]() |
Novus Biologicals
NBP227401SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP227401
![]() |
Novus Biologicals
NBP227401 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HMGB1/HMG-1 | |
0.5 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators | |
P09429 | |
3146 | |
Full-length recombinant human HMGB1 was used as the immunogen for the antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
19N15F4 | |
Western Blot 1-3ug/ml∼, Flow Cytometry 0.5ug/5x10^5cells∼, Immunohistochemistry, Immunohistochemistry-Paraffin 5ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
HMGB1 | |
IgG1 κ | |
Protein G purified | |
This antibody recognizes both the box A domain (amino acids 9-79) and full length HMGB1 protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title