Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGB1/HMG-1 Antibody (1F3), Novus Biologicals™
SDP

Catalog No. p-4968584 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225148SS 0.025 mL
NBP225148 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP225148SS Supplier Novus Biologicals Supplier No. NBP225148SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.

Specifications

Antigen HMGB1/HMG-1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF
Classification Monoclonal
Clone 1F3
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Simple Western 1:500, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, CyTOF-ready
Formulation 50% PBS, 50% glycerol with 5mM Sodium Azide
Gene Accession No. P09429
Gene Alias Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
Gene Symbols HMGB1
Host Species Mouse
Immunogen Full length recombinant human HMGB1 protein expressed in and purified form E. coli. [UniProt# P09429]
Molecular Weight of Antigen 25 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3146
Test Specificity Human, Bovine, Porcine, Mouse and Rat. Other species have not been tested.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.