Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGB1/HMG-1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | HMGB1/HMG-1 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
HMGB1/HMG-1 Polyclonal specifically detects HMGB1/HMG-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HMGB1/HMG-1 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
HMGB1 | |
IgG | |
Affinity Purified | |
24 kDa |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators | |
PBS with 0.02% Sodium Azide | |
3146 | |
Antibody was raised against a 19 amino acid peptide near the center of human HMGB1. The immunogen is located within amino acids 70 - 120 of HMGB1. Amino Acid Squence: KGETKKKFKDPNAPKRPPS | |
Primary | |
HMGB1 antibody is human, mouse and rat reactive. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title