Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGB1/HMG-1 Antibody (J2E1), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP107087AF532
Description
HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
HMGB1/HMG-1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
HMGB1 | |
Human HMGB1/HMG-1 (1-215aa) purified from Baculovirus. | |
Primary | |
Human | |
IgG2b κ |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
J2E1 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
Mouse | |
0.1 mL | |
3146 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction