Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGB1/HMG-1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $402.00

Specifications

Antigen HMGB1/HMG-1
Concentration 0.5 mg/mL
Dilution Western Blot 5-10ug/ml, Simple Western 1:200
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227169SS
SDP
View Documents
Novus Biologicals
NBP227169SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP227169
SDP
View Documents
Novus Biologicals
NBP227169
0.1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

HMGB1/HMG-1 Polyclonal specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Western Blot, Simple Western.
Specifications

Specifications

HMGB1/HMG-1
Western Blot 5-10ug/ml, Simple Western 1:200
Polyclonal
Purified
RUO
Human
Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein
HMGB1
IgG
Protein A purified
The HMGB1 amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, chicken, and monkey. This antibody recognizes both A box domain (amino acids 9-79) and full length HMGB1 protein.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators
0.2 ml sterile PBS with 0.05% Sodium Azide
3146
A portion of amino acids 25-94 of human HMGB1 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.