Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGB1/HMG-1 Rabbit anti-Human, Mouse, Rat, Clone: 1C4T5, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | HMGB1/HMG-1 |
---|---|
Clone | 1C4T5 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
Classification | Monoclonal |
Description
HMGB1/HMG-1 Monoclonal antibody specifically detects HMGB1/HMG-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDownSpecifications
HMGB1/HMG-1 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human HMGB1/HMG-1 (P09429). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
1C4T5 | |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
Unconjugated | |
Rabbit | |
Cellular Markers, Chromatin Research, Neuroscience | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3146 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title