Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGB1/HMG-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$182.50 - $504.00
Specifications
Antigen | HMGB1/HMG-1 |
---|---|
Dilution | Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Knockout Validated |
Applications | Western Blot, Immunofluorescence, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
HMGB1/HMG-1 Polyclonal antibody specifically detects HMGB1/HMG-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, KnockDownSpecifications
HMGB1/HMG-1 | |
Western Blot, Immunofluorescence, KnockDown | |
Unconjugated | |
Rabbit | |
Cellular Markers, Chromatin Research, Neuroscience | |
PBS, 50% glycerol, pH7.3 | |
3146 | |
IgG | |
Affinity purified |
Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Knockout Validated | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
A synthetic acetylated peptide around K29 of human HMGB1. | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title