Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HMGB1/HMGB2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578005
Description
Positive Control: 293T, A431, HeLa, HepG2, A375, mouse brain, rat brain Predicted Reactivity: Xenopus laevis (87%), Pig (99%), Chicken (92%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HMGB1/HMGB2 encodes a member of the non-histone chromosomal high mobility group protein family. The proteins of this family are chromatin-associated and ubiquitously distributed in the nucleus of higher eukaryotic cells. In vitro studies have demonstrated that this protein is able to efficiently bend DNA and form DNA circles. These studies suggest a role in facilitating cooperative interactions between cis-acting proteins by promoting DNA flexibility. This protein was also reported to be involved in the final ligation step in DNA end-joining processes of DNA double-strand breaks repair and V(D)J recombination.
Specifications
HMGB1/HMGB2 | |
Polyclonal | |
Unconjugated | |
HMGB1 | |
Ac2-008; amphoterin; C80539; DEF; heparin-binding protein p30; high mobility group 1; high mobility group box 1; high mobility group box 2; high mobility group protein 1; High mobility group protein 2; high mobility group protein B1; High mobility group protein B2; high-mobility group (nonhistone chromosomal) protein 1; high-mobility group (nonhistone chromosomal) protein 2; high-mobility group box 1; high-mobility group box 2; HMG1; HMG-1; HMG2; HMG-2; HMG3; Hmgb1; Hmgb2; p30; SBP-1; Sulfoglucuronyl carbohydrate binding protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15289, 25459, 29395, 3146, 3148, 97165 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P09429, P26583, P30681, P52925, P63158, P63159 | |
HMGB1, HMGB2 | |
Full length human HMGB1/HMGB2 Recombinant protein. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction