Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMGB1 Monoclonal Antibody (HAP46.5)
GREENER_CHOICE

Catalog No. MA120338
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA120338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA120338 Supplier Invitrogen™ Supplier No. MA120338
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: MCF7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

High-mobility group box 1 protein, previously known as HMG-1 or amphoterin, is a member of the high mobility group box family of non-histone chromosomal proteins. Human HMGB1 is expressed as a 30 kDa, 215 amino acid (aa) single chain polypeptide containing three domains: two N-terminal globular, 70 aa positively charged DNA-binding domains (HMG boxes A and B), and a negatively charged 30 aa C-terminal region that contains only Asp and Glu. HMGB1 is expressed at high levels in almost all cells. It was originally discovered as a nuclear protein that could bend DNA. Such bending stabilizes nucleosome formation and regulates the expression of select genes upon recruitment by DNA binding proteins. It is now known that HMGB1 can also act extracellularly, both as an inflammatory mediator that promotes monocyte migration and cytokine secretion, and as a mediator of T cell-dendritic cell interaction. Moreover, HMGB1 is reported that the level of HMGB1 is elevated during sterile tissue injury, infection, lethal endotoxemia or sepsis, collagen-induced arthritis, and ischemia-reperfusion induced tissue injury.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGB1
Applications ELISA, Western Blot
Classification Monoclonal
Clone HAP46.5
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Hmgb1
Gene Accession No. P09429, P63158, P63159, Q6YKA4, Q9YH06
Gene Alias Ac2-008; amphoterin; Amphoterin antibody; DEF; DKFZp686A04236; Heparin-binding protein p30; high mobility group 1; high mobility group 1 protein; high mobility group box 1; high mobility group protein 1; High mobility group protein B1; high mobility group protein HMG1; high-mobility group (nonhistone chromosomal) protein 1; high-mobility group box 1; high-mobility-group protein; Hmg1; HMG-1; HMG3; HMG3 antibody; hmgb 1; HMGB1; hmgb-1; HMGB1 protein; hypothetical protein; non-histone protein HMG1; p30; RCJMB04_15a21; RP11-550P23.1; SBP-1; Sulfoglucuronyl carbohydrate binding protein; unnamed protein product
Gene Symbols Hmgb1
Host Species Mouse
Immunogen Purified recombinant human HMG-1.
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15289, 25459, 3146, 395724, 403170
Target Species Human, Mouse, Rat, Canine, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.