Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMGB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595367
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595367 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595367 Supplier Invitrogen™ Supplier No. PA595367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Jurkat whole cell, rat PC-12 whole cell, mouse spleen tissue. IHC: mouse intestine tissue, rat spleen tissue, human intestinal cancer tissue. ICC/IF: U20S cell, MCF-7 cell. Flow: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

HMGB2 encodes a member of the non-histone chromosomal high mobility group protein family. The proteins of this family are chromatin-associated and ubiquitously distributed in the nucleus of higher eukaryotic cells. In vitro studies have demonstrated that this protein is able to efficiently bend DNA and form DNA circles. These studies suggest a role in facilitating cooperative interactions between cis-acting proteins by promoting DNA flexibility. This protein was also reported to be involved in the final ligation step in DNA end-joining processes of DNA double-strand breaks repair and V(D)J recombination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGB2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene Hmgb2
Gene Accession No. P26583, P30681, P52925
Gene Alias C80539; high mobility group box 2; high mobility group protein 2; High mobility group protein B2; high-mobility group (nonhistone chromosomal) protein 2; high-mobility group box 2; HMG 2; HMG B2; Hmg2; HMG-2; HMGB2; HMG-B2; HMGB-2
Gene Symbols Hmgb2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human HMGB2 (65-97aa KSDKARYDREMKNYVPPKGDKKGKKKDPNAPKR).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29395, 3148, 97165
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.