Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGB3/HMG4 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298886MFV500
Description
HMGB3/HMG4 Polyclonal antibody specifically detects HMGB3/HMG4 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
HMGB3/HMG4 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
High mobility group protein 2a, High mobility group protein 4, high mobility group protein B3, high-mobility group (nonhistone chromosomal) protein 4, high-mobility group box 3, HMG-2a, HMG2AHMG-4, HMG4MGC90319, non-histone chromosomal protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human HMGB3/HMG4. | |
0.1 mL | |
Chromatin Research, DNA Repair | |
3149 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction