Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMGB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523362
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523362 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523362 Supplier Invitrogen™ Supplier No. PA523362
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HMGB3 Polyclonal Antibody for Western Blot

HMGB3 belongs to the high mobility group protein superfamily. Like HMG1 and HMG2, HMGB3 contains DNA-binding HMG box domains and is classified into the HMG box subfamily. Members of the HMG box subfamily are thought to play a fundamental role in DNA replication, nucleosome assembly and transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGB3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA and 0.05% sodium azide
Gene HMGB3
Gene Accession No. O15347, O54879, P40618, Q32L31
Gene Alias high mobility group box 3; high mobility group protein 1; high mobility group protein 2a; High mobility group protein 4; high mobility group protein B1; high mobility group protein B3; high mobilty group protein 2A; high-mobility group (nonhistone chromosomal) protein 4; high-mobility group protein 4; HMG-1; HMG2a; HMG-2a; HMG4; HMG-4; HMGB1; Hmgb3; HMGB3 transcript variant 1/2; MGC90319; RGD1564407; Unknown (protein for MGC:133786)
Gene Symbols HMGB3
Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human HMGB3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100069284, 15354, 3149, 396232, 481073, 613729
Target Species Human, Mouse, Canine, Monkey, Bovine, Horse, Chicken
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.