Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMGB4 Monoclonal Antibody (OTI5G3)
GREENER_CHOICE

Catalog No. PIMA527031
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527031 Supplier Invitrogen™ Supplier No. MA527031
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HMGB4 Monoclonal Antibody for Western Blot, IHC (P)

HMGB4 contains two HMG-box regions, which is found in a variety of eukaryotic chromosomal proteins and transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGB4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene HMGB4
Gene Accession No. Q8WW32
Gene Alias 1700001F22Rik; AI326135; dJ1007G16.5; high mobility group box 4; high mobility group protein B4; high-mobility group box 4; HMG2 like; Hmgb4; LOC539195 protein
Gene Symbols HMGB4
Host Species Mouse
Immunogen Full length human recombinant protein of HMGB4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127540
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.