Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMGB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557370
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557370 Supplier Invitrogen™ Supplier No. PA557370
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPSSFLLFCQ DHYAQLKREN PNWSVVQVAK ATGKMWSTAT DLEKHPYEQR VALLRAKYFE ELELYRKQCN ARKKYRMS Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 63%.

HMGB4 contains two HMG-box regions, which is found in a variety of eukaryotic chromosomal proteins and transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGB4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HMGB4
Gene Accession No. Q8WW32
Gene Alias 1700001F22Rik; AI326135; dJ1007G16.5; high mobility group box 4; high mobility group protein B4; high-mobility group box 4; HMG2 like; Hmgb4; LOC539195 protein
Gene Symbols HMGB4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HMGB4. Recombinant protein control fragment (Product #RP-94470).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127540
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.