Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMGB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579375
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579375 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579375 Supplier Invitrogen™ Supplier No. PA579375
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Rat Testis Tissue, JURKAT Cell, HT1080 Cell.

HMGB4 contains two HMG-box regions, which is found in a variety of eukaryotic chromosomal proteins and transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGB4
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene HMGB4
Gene Accession No. Q6P8W9, Q8WW32
Gene Alias 1700001F22Rik; AI326135; dJ1007G16.5; high mobility group box 4; high mobility group protein B4; high-mobility group box 4; HMG2 like; Hmgb4; LOC539195 protein
Gene Symbols HMGB4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human HMGB4 (24-41aa RNKFKEQQPNTYVGFKEF).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127540, 685271, 69317
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.