Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGCL Antibody (22H11), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | HMGCL |
---|---|
Clone | 22H11 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
HMGCL Monoclonal antibody specifically detects HMGCL in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
HMGCL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
3-hydroxy-3-methylglutaryl-CoA lyase, 3-hydroxymethyl-3-methylglutaryl-CoA lyase, EC 4.1.3.4, HLmitochondrial | |
A synthesized peptide derived from Human HMGCL [UniProt P35914] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
22H11 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
3155 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title