Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGCLL1 Polyclonal Antibody
HMGCLL1 Polyclonal Antibody
Supplier: Thermo Scientific PA570202
Description
HMGCLL1 Polyclonal Antibody for Western Blot

Specifications
HMGCLL1 | |
Polyclonal | |
Unconjugated | |
HMGCLL1 | |
3-hydroxy-3-methylglutaryl-CoA lyase, cytoplasmic; 3-hydroxy-3-methylglutaryl-CoA lyase-like protein 1; 3-hydroxymethyl-3-methylglutaryl-CoA lyase like 1; 3-hydroxymethyl-3-methylglutaryl-CoA lyase, cytoplasmic; 3-hydroxymethyl-3-methylglutaryl-CoA lyase-like 1; 3-hydroxymethyl-3-methylglutaryl-CoA lyase-like protein 1; 3-hydroxymethyl-3-methylglutaryl-Coenzyme A lyase-like 1; bA418P12.1; BC037381; Endoplasmic reticulum 3-hydroxy-3-methylglutaryl-CoA lyase; endoplasmic reticulum 3-hydroxymethyl-3-methylglutaryl-CoA lyase; er-cHL; HMGCLL1; probable 3-hydroxymethyl-3-methylglutaryl-CoA lyase 2; RGD1565090 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
54511 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TB92 | |
HMGCLL1 | |
synthetic peptide directed towards the N terminal region of human HMGCLL1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction