Learn More
Invitrogen™ HMGCR Monoclonal Antibody (CL0260)

Description
Immunogen sequence: GAKFLSDAEI IQLVNAKHIP AYKLETLMET HERGVSIRRQ LLSKKLSEPS SLQYLPYRDY NYSLVMGACC ENVIGYMPIP VGVAGPLCLD EKEFQVPMAT TEGCLVASTN RGCRAIGLGG GASSRVLADG MTRGPVVRLP RACDSAEVKA WLETSEGFAV IKEAFDSTSR FARLQKLHTS IAGRNLYIRF QSRSGDAMGM NMISKGTEKA LSKLHEYFPE MQILAVSGNY CTDKKPAAIN WIEGRGKSVV CEAVIPAKVV REVLKTTT Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 92%.
Specifications
Specifications
| Antigen | HMGCR |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | CL0260 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | HMGCR |
| Gene Accession No. | P04035 |
| Gene Alias | 3H3M; 3-hydroxy-3-methylglutaryl CoA reductase (NADPH); 3-hydroxy-3-methylglutaryl-CoA reductase; 3-hydroxy-3-methylglutaryl-Coenzyme A reductase; HMDH; HMG CoA reductase; HMG CoAR; HMG-CoA reductase; HMG-CoAR; HMGCR; hydroxymethylglutaryl-CoA reductase; LDLCQ3; MGC103269; Red |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.