Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HMGCR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552547
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552547 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552547 Supplier Invitrogen™ Supplier No. PA552547
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAGSIGGYNA HAANIVTAIY IACGQDAAQN VGSSNCITLM EASGPTNEDL YISCTMPSIE IGTVGGGTNL LPQQACLQML GVQGACKDNP GENARQLARI VCGTVMAGEL SLMAALAAGH LVKSHMIHNR SKINLQDLQG Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

3-Hydroxy-3-Methylglutaryl-CoA Reductase (HMGCR) is a key enzyme in the mevalonate pathway, which is critical for the biosynthesis of cholesterol and other isoprenoids. HMGCR catalyzes the conversion of HMG-CoA to mevalonate, a rate-limiting step in the production of cholesterol. Due to its central role in cholesterol synthesis, HMGCR is a primary target for statin drugs, which are used to lower cholesterol levels and reduce the risk of cardiovascular disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGCR
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HMGCR
Gene Accession No. P04035
Gene Alias 3H3M; 3-hydroxy-3-methylglutaryl CoA reductase (NADPH); 3-hydroxy-3-methylglutaryl-CoA reductase; 3-hydroxy-3-methylglutaryl-Coenzyme A reductase; HMDH; HMG CoA reductase; HMG CoAR; HMG-CoA reductase; HMG-CoAR; HMGCR; hydroxymethylglutaryl-CoA reductase; LDLCQ3; MGC103269; Red
Gene Symbols HMGCR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HMGCR. Recombinant protein control fragment (Product #RP-102327).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3156
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.