Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HMGCR Rabbit anti-Human, Clone: 7C5, Bioss
SDP

Catalog No. 501989704
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9704 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9704 Supplier Bioss Supplier No. BSM52822R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. Grik2, also known as glutamate receptor 6, belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. Recent reports have suggested that defects in the Grik2 protein may be associated with autosomal recessive mental retardation and possibly other neurological disorders such as schizophrenia. Numerous isoforms of Grik2 are known to exist and may be subject to RNA editing within the second transmembrane domain, which is thought to alter the properties of ion flow. This Grik2 antibody may exhibit some cross-reactivity to Grik3.

Specifications

Antigen HMGCR
Applications Western Blot
Classification Monoclonal
Clone 7C5
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH
Gene HMGCR
Gene Accession No. P04035
Gene Alias 3H3M; 3-hydroxy-3-methylglutaryl CoA reductase (NADPH); 3-hydroxy-3-methylglutaryl-CoA reductase; 3-hydroxy-3-methylglutaryl-coenzyme A reductase; HMDH; HMG CoA reductase; HMG CoAR; HMG-CoA reductase; HMG-CoAR; HMGCR; hydroxymethylglutaryl-CoA reductase; LDLCQ3; MGC103269; Red
Gene Symbols HMGCR
Host Species Rabbit
Immunogen Human HMGCR aa 400-500.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3156
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.