Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HMGCR Recombinant Rabbit Monoclonal Antibody (6Y9L8)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535242
Description
Immunogen sequence: IKPLVAETDT PNRATFVVGN SSLLDTSSVL VTQEPEIELP REPRPNEECL QILGNAEKGA KFLSDAEIIQ LVNAKHIPAY KLETLMETHE RGVSIRRQLL S.
3-Hydroxy-3-Methylglutaryl-CoA Reductase (HMGCR) is a key enzyme in the mevalonate pathway, which is critical for the biosynthesis of cholesterol and other isoprenoids. HMGCR catalyzes the conversion of HMG-CoA to mevalonate, a rate-limiting step in the production of cholesterol. Due to its central role in cholesterol synthesis, HMGCR is a primary target for statin drugs, which are used to lower cholesterol levels and reduce the risk of cardiovascular disease.
Specifications
HMGCR | |
Recombinant Monoclonal | |
0.3 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04035, P51639, Q01237 | |
HMGCR | |
A synthetic peptide corresponding to a sequence within amino acids 400-500 of human HMGCR (P04035). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
6Y9L8 | |
Unconjugated | |
HMGCR | |
3H3M; 3-hydroxy-3-methylglutaryl CoA reductase (NADPH); 3-hydroxy-3-methylglutaryl-CoA reductase; 3-hydroxy-3-methylglutaryl-Coenzyme A reductase; HMDH; HMG CoA reductase; HMG CoAR; HMG-CoA reductase; HMG-CoAR; HMGCR; hydroxymethylglutaryl-CoA reductase; LDLCQ3; MGC103269; Red | |
Rabbit | |
Affinity chromatography | |
RUO | |
15357, 25675, 3156 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction