Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HMGCS1 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297737B
Description
HMGCS1 Polyclonal specifically detects HMGCS1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
HMGCS1 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase, 3-hydroxy-3-methylglutaryl coenzyme A synthase, 3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble), 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble), EC 2.3.3.10, HMG-CoA synthase, HMGCS, hydroxymethylglutaryl-CoA synthase, cytoplasmic, MGC90332 | |
Produced in rabbits immunized with E. coli-derived Human HMGCS1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
3157 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction