Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HMGCS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA513604
Description
This antibody is predicted to react with chicken, hamster, mouse and rat based on sequence homology.
This enzyme condenses acetyl-CoA with acetoacetyl-CoA to form HMG-CoA, which is the substrate for HMG-CoA reductase.
Specifications
HMGCS1 | |
Polyclonal | |
Unconjugated | |
HMGCS1 | |
3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase; 3-hydroxy-3-methylglutaryl coenzyme A synthase; 3-hydroxy-3-methylglutaryl-CoA synthase 1; 3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1; 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble) isoform 1; 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble) isoform 2; B130032C06Rik; HMG-CoA synthase; HMGCS; Hmgcs1; HMGCS1 protein-like; hydroxymethylglutaryl-CoA synthase 1; hydroxymethylglutaryl-CoA synthase, cytoplasmic; Unknown (protein for IMAGE:7955111) | |
Rabbit | |
Ammonium Sulfate Precipitation, Size-exclusion - Dialysis | |
RUO | |
3157 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q01581 | |
HMGCS1 | |
KLH conjugated synthetic peptide between 442-470 amino acids from the C-terminal region of human HMGCS1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction