Learn More
HMGCS1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304590AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
This enzyme condenses acetyl-CoA with acetoacetyl-CoA to form HMG-CoA, which is the substrate for HMG-CoA reductase.Specifications
HMGCS1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q01581, Q8JZK9 | |
Hmgcs1 | |
Between 470 and 520 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
HMGCS1 | |
Hydroxymethylglutaryl-CoA synthase, cytoplasmic; HMG-CoA synthase; 3-hydroxy-3-methylglutaryl coenzyme A synthase; HMGCS; HMGCS1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
208715, 3157 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.